missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tris(trifluoromethanesulfonyl)methanide 98.0+%, TCI America™

Catalog No. P25205G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25205G 5 g
P25201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25205G Supplier TCI America Supplier No. P25205G

Chemical Identifiers

CAS 114395-69-6
Molecular Formula C4F9KO6S3
Molecular Weight (g/mol) 450.302
MDL Number MFCD18072174
InChI Key RJPWSGDBEHVWPP-UHFFFAOYSA-N
PubChem CID 52991969
IUPAC Name potassium;bis(trifluoromethylsulfonyl)methylsulfonyl-trifluoromethane
SMILES [C-](S(=O)(=O)C(F)(F)F)(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F.[K+]

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 450.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Tris(trifluoromethanesulfonyl)methanide