accessibility menu, dialog, popup

UserName

CSOOFFFSOOFFFSOOFFFK

CAS RN 114395-69-6

IUPAC Name: potassium ditrifluoromethanesulfonylmethanidesulfonyltrifluoromethane
Synonyms: potassium ditrifluoromethanesulfonylmethanidesulfonyltrifluoromethane
Molecular Weight (g/mol): 450.3
Molecular Formula: C4F9KO6S3
InChi Key: RJPWSGDBEHVWPP-UHFFFAOYSA-N
SMILES: [K+].FC(F)(F)S(=O)(=O)[C-](S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results