missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tris(3,5-dimethylpyrazol-1-yl)borohydride 98.0+%, TCI America™

Catalog No. p-7135139
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
P14415G 5 g
P144125G 25 g
2 options

Catalog No. P14415G

Supplier: TCI America P14415G

Only null left

Chemical Identifiers

CAS 17567-17-8
Molecular Formula C15H22BKN6
Molecular Weight (g/mol) 336.29
MDL Number MFCD00040342
InChI Key NTWZGFNSHCFHIJ-UHFFFAOYSA-N
Synonym potassium hydrotris 3,5-dimethylpyrazol-1-yl borate, potassium;tris 3,5-dimethylpyrazol-1-yl boron 1-, potassium tri 3,5-dimethyl-1-pyrazolyl borohydride, potassium tris 3,5-dimethyl-1h-pyrazol-1-yl hydrido borate 1-
IUPAC Name potassium tris(3,5-dimethyl-1H-pyrazol-1-yl)boranuide
SMILES [K+].CC1=NN([BH-](N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2813
Formula Weight 336.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Tris(3,5-dimethylpyrazol-1-yl)borohydride