Learn More
Potassium trifluoroacetate, 98%
CAS: 2923-16-2 | C2F3KO2 | 152.11 g/mol
$95.68 - $259.49
Chemical Identifiers
| CAS | 2923-16-2 |
|---|---|
| Molecular Formula | C2F3KO2 |
| Molecular Weight (g/mol) | 152.11 |
| MDL Number | MFCD00013215 |
| InChI Key | CUNPJFGIODEJLQ-UHFFFAOYSA-M |
| Synonym | potassium trifluoroacetate, potassium 2,2,2-trifluoroacetate, trifluoroacetic acid potassium salt, acetic acid, trifluoro-, potassium salt, potassiumtrifluoroacetate, pubchem12605, potassium ion trifluoroacetate, ksc492g5t, trifluoroacetic acid, potassium salt, potassium 2,2,2-tris fluoranyl ethanoate |
| PubChem CID | 23662811 |
| SMILES | [K+].[O-]C(=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1739814
|
Thermo Scientific Chemicals
A1739814 |
25 g |
Each for $95.68
|
|
|||||
|
AAA1739822
|
Thermo Scientific Chemicals
A1739822 |
100 g |
Each for $259.49
|
|
|||||
Description
It is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, acids.
Chemical Identifiers
| 2923-16-2 | |
| 152.11 | |
| CUNPJFGIODEJLQ-UHFFFAOYSA-M | |
| 23662811 |
| C2F3KO2 | |
| MFCD00013215 | |
| potassium trifluoroacetate, potassium 2,2,2-trifluoroacetate, trifluoroacetic acid potassium salt, acetic acid, trifluoro-, potassium salt, potassiumtrifluoroacetate, pubchem12605, potassium ion trifluoroacetate, ksc492g5t, trifluoroacetic acid, potassium salt, potassium 2,2,2-tris fluoranyl ethanoate | |
| [K+].[O-]C(=O)C(F)(F)F |
Specifications
| 2923-16-2 | |
| 7 | |
| MFCD00013215 | |
| UN2811 | |
| Hygroscopic | |
| Soluble in water. | |
| [K+].[O-]C(=O)C(F)(F)F | |
| 152.11 | |
| 152.12 | |
| Potassium trifluoroacetate |
| 142°C to 145°C | |
| C2F3KO2 | |
| 25 g | |
| 3717603 | |
| potassium trifluoroacetate, potassium 2,2,2-trifluoroacetate, trifluoroacetic acid potassium salt, acetic acid, trifluoro-, potassium salt, potassiumtrifluoroacetate, pubchem12605, potassium ion trifluoroacetate, ksc492g5t, trifluoroacetic acid, potassium salt, potassium 2,2,2-tris fluoranyl ethanoate | |
| CUNPJFGIODEJLQ-UHFFFAOYSA-M | |
| potassium;2,2,2-trifluoroacetate | |
| 23662811 | |
| 98% |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 220-877-5
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only