missing translation for 'onlineSavingsMsg'
Learn More

Potassium Trifluoro(trifluoromethyl)borate 98.0+%, TCI America™

Catalog No. p-7135211
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16921G 1 g
P16925G 5 g
P169225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P16921G Supplier TCI America Supplier No. P16921G
Only null left

Chemical Identifiers

CAS 42298-15-7
Molecular Formula CBF6K
Molecular Weight (g/mol) 175.91
InChI Key UOGBGCVSVMQVBR-UHFFFAOYSA-N
PubChem CID 23674658
IUPAC Name potassium;trifluoro(trifluoromethyl)boranuide
SMILES [B-](C(F)(F)F)(F)(F)F.[K+]

Specifications

Color White
Quantity 1 g
Formula Weight 175.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Trifluoro(trifluoromethyl)borate