missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tricyanomethanide 98.0+%, TCI America™

Catalog No. p-7135594
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25381G 1 g
P25385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25381G Supplier TCI America Supplier No. P25381G
Only null left

Chemical Identifiers

CAS 34171-69-2
Molecular Formula C4KN3
Molecular Weight (g/mol) 129.163
MDL Number MFCD00058850
InChI Key ZKJPYQKGNUBNOA-UHFFFAOYSA-N
Synonym potassium tricyanomethanide, potassium methanetricarbonitrile, potassium cyanoform, acmc-1ajhe, potassium ion tricyanomethanide, tricyanomethanide potassium salt, potassiomethanetricarbonitrile
PubChem CID 10964525
IUPAC Name potassium;methanetricarbonitrile
SMILES C(#N)[C-](C#N)C#N.[K+]

Specifications

Melting Point 298°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Tricyanomethanide