missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tri-sec-butylborohydride, 1M solution in tetrahydrofuran, AcroSeal™, Thermo Scientific™
Specifications
| CAS | 54575-49-4 |
|---|---|
| Molecular Formula | C12H28BK |
| Molecular Weight (g/mol) | 222.26 |
| MDL Number | MFCD00009324 |
| Synonym | k-selectride, potassium tri-sec-butylborohydride solution, k-selectride™ solution, k-selectride r solution 1.0 m potassium tri-sec-butylborohydride in thf, k-selectride r solution, 1.0 m potassium tri-sec-butylborohydride in thf, potassium tri-sec-butylborohydride, 1.0m in thf, in sure/seal tm bottle |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC205478000
|
Thermo Scientific Chemicals
205478000 |
800mL | AcroSeal™ Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagentSpecifications
| 54575-49-4 | |
| 222.26 | |
| k-selectride, potassium tri-sec-butylborohydride solution, k-selectride™ solution, k-selectride r solution 1.0 m potassium tri-sec-butylborohydride in thf, k-selectride r solution, 1.0 m potassium tri-sec-butylborohydride in thf, potassium tri-sec-butylborohydride, 1.0m in thf, in sure/seal tm bottle |
| C12H28BK | |
| MFCD00009324 |
RUO â Research Use Only