accessibility menu, dialog, popup

UserName

CAS RN 54575-49-4

BH3CH3CH3CCH3CH3H3CHK

CAS RN 54575-49-4

IUPAC Name: boron(3+) potassium tris(butan-2-ide) hydride
Synonyms: boron(3+) potassium tris(butan-2-ide) hydride
Molecular Weight (g/mol): 222.26
Molecular Formula: C12H28BK
InChi Key: UHNDOQOAVCJJPO-UHFFFAOYSA-N
SMILES: [H-].[B+3].[K+].CC[CH-]C.CC[CH-]C.CC[CH-]C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)