missing translation for 'onlineSavingsMsg'
Learn More

Potassium tin(IV) oxide trihydrate, 95%

Catalog No. p-7049328
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1401530 250 g
AA14015A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1401530 Supplier Thermo Scientific Chemicals Supplier No. 01401530
Only null left

CAS: 12125-03-0 | H6K2O6Sn | 298.95 g/mol

Potassium tin(IV) oxide is a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12125-03-0
Molecular Formula H6K2O6Sn
Molecular Weight (g/mol) 298.95
MDL Number MFCD00150391
InChI Key HTHDWDSBYOUAFF-UHFFFAOYSA-N
Synonym potassium stannate trihydrate, unii-r38y86o4sy, 2k.so3n.3h2o, potassium tin iv oxide trihydrate, dipotassium dioxido oxo tin trihydrate, dipotassium ion oxostannanebis olate trihydrate, dipotassium oxostannanebis olate trihydrate, dipotassium bis oxidanidyl-oxidanylidene-tin trihydrate
PubChem CID 18694711
IUPAC Name dipotassium;dioxido(oxo)tin;trihydrate
SMILES O.O.O.[K+].[K+].[O-][Sn]([O-])=O

Specifications

Melting Point 140°C (decomposition)
Physical Form Crystalline
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 298.94 (244.89 Anhydrous)
Percent Purity ≥95%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium tin(IV) oxide trihydrate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only