missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog No. 60045940
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
6 x 250 g
6 x 1 kg
6 x 2.5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045940 6 x 250 g
6002344 250 g
6002342 1 kg
6002343 2.5 kg
60045938 6 x 1 kg
60045939 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60045940 Supplier Honeywell Chemicals Supplier No. 312726X250G
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Quantity 6 x 250 g
Linear Formula KSCN
UN Number NONH for all modes of transport
Formula Weight 97.18
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Thiocyanate, Reag. Ph. Eur. reag. ISO
Hazard Statement P273-P280
Precautionary Statement H302 + H312 + H332-H412
EINECSNumber 206-370-1