missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Purified, Spectrum™ Chemical

Catalog No. 18612936
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612936 50 kg Poly Bottle
18612933 500 g Poly Bottle
18612934 2.5 kg Poly Bottle
18612935 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612936 Supplier Spectrum Chemical Mfg Cor Supplier No. P142150KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 333-20-0

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Grade Purified
Packaging Poly Bottle
Chemical Name or Material Potassium Thiocyanate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.