missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. P317100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P317100 100 g Glass Bottle
P317500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P317100 Supplier Thermo Fisher Scientific Supplier No. P317 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 170°C
Color White
pH 5.3 to 8.7
Physical Form Crystalline Solid
Quantity 100 g
Assay Percent Range ≥98.5 %
Formula Weight 97.18
Percent Purity ≥98.5%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure <1hPa at 20°C
Density 1.886g/cm³
Chemical Name or Material Potassium Thiocyanate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Contact with acids liberates very toxic gas. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:2
Flammability:0
Instability:1