missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate concentrate, For 1L standard solution, 0.1 M KSCN, Honeywell Fluka™

Catalog No. p-7122106
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002670 1 Pc.
60046699 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002670 Supplier Honeywell Chemicals Supplier No. 381401EA
Only null left

for 1L standard solution, 0.1 M KSCN

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Quantity 1 Pc.
Linear Formula KSCN
UN Number NONH for all modes of transport
Beilstein 3594799
Formula Weight 97.18g/mol
Packaging Ampoule
Density 1g/mL at 25°C
Chemical Name or Material Potassium thiocyanate concentrate