missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Certified, 10.0% (w/v) ±0.4%, LabChem™

Supplier:  LabChem LC221402

 View more versions of this product

Catalog No. LC221402


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Thiocyanate
Colorless
Liquid
CKNS
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium;thiocyanate
516872
97.18
Certified
Poly Bottle
333-20-0,7732-18-5
90,10
1 L
KSCN
Soluble in water
C(#N)[S-].[K+]
97.176
CHEBI:30951
10.0% ±0.4% (w/v)
Passes Test
Hydrogen cyanide; Nitrogen oxides; Sulfur compounds; Carbon monoxide; Carbon dioxide

Chemical Identifiers

333-20-0
97.176
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
CHEBI:30951
C(#N)[S-].[K+]
CKNS
ZNNZYHKDIALBAK-UHFFFAOYSA-M
516872
potassium;thiocyanate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions