missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
Supplier: LabChem LC221702
Specifications
| Potassium Thiocyanate | |
| Colorless | |
| Liquid | |
| CKNS | |
| potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
| ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
| potassium;thiocyanate | |
| 516872 | |
| 97.18 | |
| 0.1000N ±0.0005N (0.1M) | |
| Poly Bottle |
| 333-20-0,7732-18-5 | |
| 99.03,0.97 | |
| 1 L | |
| KSCN | |
| Soluble in water | |
| C(#N)[S-].[K+] | |
| 97.176 | |
| CHEBI:30951 | |
| Certified | |
| Passes Test | |
| Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds |
Chemical Identifiers
| 333-20-0 | |
| 97.176 | |
| potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
| CHEBI:30951 | |
| C(#N)[S-].[K+] |
| CKNS | |
| ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
| 516872 | |
| potassium;thiocyanate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature