missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCP0730100
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
10 kg
Packaging:
Fiber Drum
White Poly Wide Mouth Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCP0730100 100 g White Poly Wide Mouth Bottle
RDCP0730500 500 g White Poly Wide Mouth Bottle
RDCP073010F 10 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. RDCP0730100 missing translation for 'mfr' Ricca Chemical Company missing translation for 'supplierNo' RDCP0730100B1

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
PubChem CID 516872
ChEBI CHEBI:30951

Specifications

Color Colorless or White
CAS Min % 98.0
pH 5.3 to 8.7
CAS Max % 100.0
Physical Form Crystals
Quantity 100 g
Synonym potassium thiocyanate
SMILES [K+].[S-]C#N
IUPAC Name potassium cyanosulfanide
Molecular Weight (g/mol) 97.18
Percent Purity ≥98.5%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Appearance Colorless/White Solid
Chemical Name or Material Potassium, Thiocyanate
Show More Show Less
ShelfLife 60 months