missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, ACS Grade, ≥ 98.5%, LabChem™

Supplier:  LabChem LC201201

Catalog No. LC201201


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Thiocyanate
White
Crystalline
CKNS
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium;thiocyanate
516872
97.18
ACS
Poly Bottle
333-20-0
100
500 g
KSCN
Soluble in water
C(#N)[S-].[K+]
97.176
CHEBI:30951
≥98.5%
Passes Test
Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds

Chemical Identifiers

333-20-0
97.176
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
CHEBI:30951
C(#N)[S-].[K+]
CKNS
ZNNZYHKDIALBAK-UHFFFAOYSA-M
516872
potassium;thiocyanate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions