missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 99%, ACS reagent

Catalog No. AC424235000
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424235000 500 g Plastic bottle
AC424230050 5 g Glass bottle
AC424230025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424235000 Supplier Thermo Scientific Chemicals Supplier No. 424235000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 333-20-0 | CKNS | 97.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Color Colorless or White
pH 5.3 to 8.7 (5% soln. at 25°C)
Physical Form Crystals
Quantity 500 g
Packaging Plastic bottle
Vapor Pressure <1hPa at 20°C
Assay Percent Range 99%
Density 1.886g/mL
Linear Formula KSCN
Fieser 01,954
Merck Index 15, 7807
Molecular Weight (g/mol) 97.176
Formula Weight 97.18
Percent Purity ≥98.5%
Grade ACS Reagent
Chemical Name or Material Potassium thiocyanate
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-370-1
RTECSNumber XL1925000
TSCA TSCA

RUO – Research Use Only