missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 98%

Catalog No. AAA1373136
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1373136 500 g
AAA137310E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1373136 Supplier Thermo Scientific Chemicals Supplier No. A1373136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 333-20-0 | CKNS | 97.176 g/mol

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

Solubility
Soluble in water, acetone and alcohol.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with nitric acids, active halogen compounds and strong bases.

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 172°C to 175°C
Density 1.89
pH 5.3 to 8.5
Boiling Point 500°C (decomposition)
Odor Odorless
Quantity 500 g
Beilstein 3594799
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,7691
Solubility Information Soluble in water,acetone and alcohol.
Formula Weight 97.18
Percent Purity 98%
Chemical Name or Material Potassium thiocyanate
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 206-370-1
RTECSNumber XL1925000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only