missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, 4% (w/v), Ricca Chemical
APHA for Iodide Testing
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6550-16
|
Ricca Chemical Company
655016 |
500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
6550-32
|
Ricca Chemical Company
655032 |
1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| Potassium Thiocyanate | |
| APHA for Iodide |
Specifications
| 500 mL | |
| 500 mL | |
| Potassium Thiocyanate |
| Natural Poly Bottle | |
| APHA for Iodide |