missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, 4% (w/v), Ricca Chemical
APHA for Iodide Testing
Supplier: Ricca Chemical Company 655016
Specifications
| 500 mL | |
| 500 mL | |
| Potassium Thiocyanate |
| Natural Poly Bottle | |
| APHA for Iodide |
Chemical Identifiers
| Potassium Thiocyanate | |
| APHA for Iodide |