Learn More
Potassium tetrafluoroborate, 99%, extra pure
CAS: 14075-53-7 | BF4K | 125.90 g/mol
Supplier: Thermo Scientific Chemicals 269220010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
| Potassium tetrafluoroborate | |
| c.p. | |
| White to Gray | |
| 1 kg | |
| BF4K | |
| MFCD00011395 | |
| potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride | |
| AKEBROIVCDHVSD-UHFFFAOYSA-N | |
| potassium tetrafluoroboranuide | |
| 518872 | |
| 99% | |
| Plastic bottle |
| 530.0°C | |
| 14075-53-7 | |
| Micro-crystalline Powder | |
| 98.5% min | |
| KBF4 | |
| 15,7804 | |
| Solubility in water: 4.4g/L (20°C). Other solubilities: slightly soluble in boiling alcohol | |
| [K+].F[B-](F)(F)F | |
| 125.90 | |
| 125.91 | |
| Extra Pure | |
| 2.5 |
Chemical Identifiers
| 14075-53-7 | |
| 125.90 | |
| AKEBROIVCDHVSD-UHFFFAOYSA-N | |
| 518872 | |
| [K+].F[B-](F)(F)F |
| BF4K | |
| MFCD00011395 | |
| potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride | |
| potassium tetrafluoroboranuide |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 237-928-2
RTECSNumber : ED2800000
TSCA : TSCA
RUO – Research Use Only