missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetracyanonickelate(II) hydrate
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C (in -H2O) |
| Physical Form | Crystalline |
| Quantity | 10 g |
| Density | 1.875g/mL |
| UN Number | UN1588 |
| Merck Index | 14,7684 |
| Solubility Information | Soluble in water |
| Formula Weight | 240.99 (Anhydrous) |
| Chemical Name or Material | Potassium tetracyanonickelate(II) hydrate |
Safety and Handling
| Hazard Category | H300+H310+H330-H317-H334-H350i |
| Hazard Statement | GHS H Statement H301-H334-H350-H317 Toxic if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P262-P264b-P270-P271-P272-P280g-P281-P284-P285-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P333+P313-P361-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 238-082-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only