accessibility menu, dialog, popup

UserName

KKNiNNNN

CAS RN 339527-86-5

IUPAC Name: dipotassium tetracyanonickeldiuide
Synonyms: dipotassium tetracyanonickelate(2-)
Molecular Weight (g/mol): 240.96
Molecular Formula: C4K2N4Ni
InChi Key: QOOHKAMDWOKKBR-UHFFFAOYSA-N
SMILES: [K+].[K+].N#C[Ni--](C#N)(C#N)C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results