missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, 98+%, pure

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

Supplier:  Thermo Scientific Chemicals 168880250

 View more versions of this product

Catalog No. AC168880250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium tert-butoxide
240.0°C to 258.0°C
98+%
C4H9KO
25 g
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
23665647
98+%
Powder
865-47-4
White
Plastic bottle
(CH3)3COK
01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Solubility in water: reacts with water. Other solubilities: soluble in methanol, ethanol, slightly soluble in diethyl ether, benzene
CC(C)(C)[O-].[K+]
112.21
112.21
Pure

Chemical Identifiers

865-47-4
112.21
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
potassium;2-methylpropan-2-olate
C4H9KO
LPNYRYFBWFDTMA-UHFFFAOYSA-N
23665647
CC(C)(C)[O-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Self-heating in large quantities; may catch fire.
Reacts violently with water.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several min

GHS Signal Word: Danger

EINECSNumber : 212-740-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only