missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-Butoxide (12% in Tetrahydrofuran, ca. 1mol/L), TCI America™

Catalog No. p-7135179
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1619100ML 100 mL
P1619500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1619100ML Supplier TCI America Supplier No. P1619100ML
Only null left

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.213
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]

Specifications

Color Yellow
Quantity 100 mL
UN Number 2924
Formula Weight 112.21
Physical Form Liquid
Chemical Name or Material Potassium tert-Butoxide (12% in Tetrahydrofuran, ca. 1mol/L)
EINECSNumber (2)-3100
TSCA Yes