missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfite, 90%, pure

Catalog No. ac440212500
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440212500 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440212500 Supplier Thermo Scientific Chemicals Supplier No. 440212500
Only null left

CAS: 10117-38-1 | K2O3S | 158.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10117-38-1
Molecular Formula K2O3S
Molecular Weight (g/mol) 158.26
InChI Key BHZRJJOHZFYXTO-UHFFFAOYSA-L
Synonym potassium sulfite, dipotassium sulfite, potassium sulphite, sulfurous acid, dipotassium salt, unii-015kzc652e, sulfurous acid, potassium salt 1:2, potassium sulfite k2so3, kaliumsulfit, potassiumsulfite, dipotassium ion sulfite
PubChem CID 24958
IUPAC Name dipotassium;sulfite
SMILES [O-]S(=O)[O-].[K+].[K+]

Specifications

Quantity 250 g
Assay Percent Range 88% min. (Iodimetry)
Linear Formula K2SO3
Merck Index 15,7795
Formula Weight 158.26
Percent Purity 90%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Potassium sulfite
EINECSNumber 233-321-1

RUO – Research Use Only