accessibility menu, dialog, popup

UserName

KKOSOO

CAS RN 10117-38-1

IUPAC Name: dipotassium sulfite
Synonyms: potassium sulfite
Molecular Weight (g/mol): 158.25
Molecular Formula: K2O3S
InChi Key: BHZRJJOHZFYXTO-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]S([O-])=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results