missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99%, Honeywell Fluka™

Catalog No. p-7072018
Change view
Click to view available options
Quantity:
1 kg
5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002340 1 kg
6002341 5 kg
60045937 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6002340 Supplier Honeywell Chemicals Supplier No. 312701KG
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula K2SO4
UN Number NONH for all modes of transport
IUPAC Name dipotassium;sulfate
Formula Weight 174.26g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Sulfate, Reag. ISO
EINECSNumber 231-915-5
RTECSNumber TT5900000