missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1689.0°C |
| Melting Point | 1067.0°C |
| Color | White |
| pH | 5.5 to 8.5 |
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Linear Formula | K2SO4 |
| Merck Index | 15,7793 |
| Solubility Information | Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol |
| IUPAC Name | dipotassium sulfate |
| Show More |
Safety and Handling
| EINECSNumber | 231-915-5 |
RUO – Research Use Only