missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, meets analytical spec. of Ph. Eur., 99-101%, Honeywell Fluka™
Meets analytical specification of Ph. Eur., 99-101%
$323.38 - $323.38
Chemical Identifiers
| CAS | 7778-80-5 |
|---|---|
| Molecular Formula | K2O4S |
| Molecular Weight (g/mol) | 174.25 |
| MDL Number | MFCD00011388 |
| InChI Key | OTYBMLCTZGSZBG-UHFFFAOYSA-L |
| Synonym | potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt |
| PubChem CID | 24507 |
| ChEBI | CHEBI:32036 |
| SMILES | [K+].[K+].[O-]S([O-])(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7778-80-5 | |
| 174.25 | |
| OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
| 24507 | |
| [K+].[K+].[O-]S([O-])(=O)=O |
| K2O4S | |
| MFCD00011388 | |
| potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
| CHEBI:32036 |
Specifications
| 7778-80-5 | |
| K2O4S | |
| MFCD00011388 | |
| potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
| OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
| dipotassium;sulfate | |
| 24507 | |
| 174.26g/mol | |
| Poly bottle |
| 1 kg | |
| K2SO4 | |
| NONH for all modes of transport | |
| Insoluble in alcohol, Soluble in boiling water 1gm/4mL, Soluble in glycerol 1gm/75mL, Soluble in water 1gm/8.3mL | |
| [K+].[K+].[O-]S([O-])(=O)=O | |
| 174.25 | |
| CHEBI:32036 | |
| 99 to 101% | |
| Potassium Sulfate, Meets analytical specification of Ph.Eur. |
Safety and Handling
EINECSNumber : 231-915-5
RTECSNumber : TT5900000