missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, For Analysis, EMSURE™, MilliporeSigma™,

Catalog No. M1051530500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1051530500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1051530500 Supplier Technidata Supplier No. 1.05153.0500
Only null left

Recommended Storage

+5°C to +30°C.

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C (1013hPa)
Melting Point 1069°C
Color White
pH 5.5 to 8.5 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 500 g
Solubility Information 111g/L
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 2.66 g/3 (20 °C)
Chemical Name or Material Potassium Sulfate
Show More Show Less
EINECSNumber 231-915-5
RTECSNumber TT5900000
Recommended Storage +5°C to +30°C.