missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sulfate, FCC, 99-100.5%, Spectrum™ Chemical
P1589, 7778-80-5, K2SO4
$254.77 - $9299.90
Chemical Identifiers
| CAS | 7778-80-5 |
|---|---|
| Molecular Formula | K2O4S |
| Molecular Weight (g/mol) | 174.25 |
| MDL Number | MFCD00011388 |
| InChI Key | OTYBMLCTZGSZBG-UHFFFAOYSA-L |
| IUPAC Name | dipotassium sulfate |
| SMILES | [K+].[K+].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605730
|
Spectrum Chemical Mfg Cor
P1589125GM |
125 g | Poly Bottle |
Each for $254.77
|
|
||||
|
18605731
|
Spectrum Chemical Mfg Cor
P1589500GM |
500 g | Poly Bottle |
Each for $385.97
|
|
||||
|
18605732
|
Spectrum Chemical Mfg Cor
P158925KG |
2.5 kg | Poly Pail |
Each for $1,391.07
|
|
||||
|
18605733
|
Spectrum Chemical Mfg Cor
P158912KG |
12 kg | Poly Pail |
Each for $3,761.63
|
|
||||
|
18605734
|
Spectrum Chemical Mfg Cor
P158950KGBL |
50 kg | Fiber Drum |
Each for $9,299.90
|
|
||||