missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-33229
Change view
Click to view available options
Quantity:
500 g
10 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P304500 500 g Poly Bottle
P30410 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P304500 Supplier Thermo Fisher Scientific Supplier No. P304 500
Only null left

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C
Melting Point 1,067°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99 %
Formula Weight 174.26
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Potassium Sulfate
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0