missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, 99%

Catalog No. AAA139750I
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA139750I 5000 g
AAA139750B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA139750I Supplier Thermo Scientific Chemicals Supplier No. A139750I

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate and glass. In analytical chemistry, it is used in the determination of nitrogen by the Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is used in the separation of protein through electrophoresis and also serves as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate and glass. In analytical chemistry, it is used in the determination of nitrogen by the Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is used in the separation of protein through electrophoresis and also serves as a laboratory reagent.

Solubility
Soluble in water and glycerol. Insoluble in alcohol, acetate and carbon disulfide.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C
Melting Point 1,069°C
Quantity 5000 g
Merck Index 14,7674
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide.
Formula Weight 174.27
Percent Purity 99%
Odor Odorless
Density 2.662
Chemical Name or Material Potassium Sulfate
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-915-5
RTECSNumber TT5900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only