missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium sulfate, 99+%, for biochemistry

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

$154.44 - $154.44

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC424212500
SDS View Documents
Thermo Scientific Chemicals
424212500
250 g Plastic Bottle
Each for $154.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7778-80-5
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
dipotassium sulfate
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
CHEBI:32036
[K+].[K+].[O-]S([O-])(=O)=O

Specifications

1689.0°C
7778-80-5
Crystals
99+%
K2SO4
15,7793
(10 g in 150 ml water) Clear colorless
[K+].[K+].[O-]S([O-])(=O)=O
174.25
CHEBI:32036
≥99%
Plastic Bottle
1067.0°C
White
250 g
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
24507
174.25
Biochemical
Potassium sulfate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-915-5

RTECSNumber : TT5900000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only