missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, 99+%, for analysis, anhydrous, granular

Catalog No. p-43501
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205945000 500 g Plastic bottle
AC205940025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC205945000 Supplier Thermo Scientific Chemicals Supplier No. 205945000
Only null left

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689.0°C
Melting Point 1067.0°C
Color White
pH 5.5 to 8.5 (5% soln. at 25°C)
Physical Form Granular Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula K2SO4
Merck Index 15, 7793
Solubility Information Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol
IUPAC Name dipotassium;sulfate
Formula Weight 174.25
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 2.66
Chemical Name or Material Potassium sulfate, p.a.
Show More Show Less
EINECSNumber 231-915-5
RTECSNumber TT5900000
TSCA TSCA

RUO – Research Use Only