missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, 99+%, ACS reagent
CAS: 7778-80-5 | K2O4S | 174.25 g/mol
Supplier: Thermo Scientific Chemicals 424220050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
| 1689.0°C | |
| 1067.0°C | |
| White | |
| Fine Crystals or Powder | |
| 99+% | |
| K2SO4 | |
| 15, 7793 | |
| Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol | |
| [K+].[K+].[O-]S([O-])(=O)=O | |
| 174.25 | |
| CHEBI:32036 | |
| ≥99% | |
| Plastic bottle |
| Potassium sulfate | |
| 7778-80-5 | |
| 5.5 to 8.5 (5% soln. at 25°C) | |
| 5 kg | |
| K2O4S | |
| MFCD00011388 | |
| potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
| OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
| dipotassium;sulfate | |
| 24507 | |
| 174.25 | |
| ACS Reagent | |
| 2.66g/mL |
Chemical Identifiers
| 7778-80-5 | |
| 174.25 | |
| OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
| 24507 | |
| [K+].[K+].[O-]S([O-])(=O)=O |
| K2O4S | |
| MFCD00011388 | |
| potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
| CHEBI:32036 |
Safety and Handling
EINECSNumber : 231-915-5
RUO – Research Use Only