missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, 99+%, ACS reagent

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

Supplier:  Thermo Scientific Chemicals 424220010

 View more versions of this product

Catalog No. AC424220010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

1689.0°C
1067.0°C
White
Fine Crystals or Powder
99+%
K2SO4
15, 7793
Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol
[K+].[K+].[O-]S([O-])(=O)=O
174.25
CHEBI:32036
≥99%
Plastic bottle
Potassium sulfate
7778-80-5
5.5 to 8.5 (5% soln. at 25°C)
1 kg
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
24507
174.25
ACS Reagent
2.66g/mL

Chemical Identifiers

7778-80-5
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
[K+].[K+].[O-]S([O-])(=O)=O
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
CHEBI:32036
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-915-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only