missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, 99+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Boiling Point | 1689.0°C |
| Melting Point | 1067.0°C |
| Color | White |
| pH | 5.5 to 8.5 (5% soln. at 25°C) |
| Physical Form | Fine Crystals or Powder |
| Quantity | 250 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 99+% |
| Density | 2.66g/mL |
| Linear Formula | K2SO4 |
| Show More |
Safety and Handling
| EINECSNumber | 231-915-5 |
RUO – Research Use Only