missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sorbate, Powder, FCC, 98-101%, Spectrum™ Chemical
P1408, 590-00-1, C6H7KO2
$327.13 - $13243.77
Chemical Identifiers
| CAS | 590-00-1 |
|---|---|
| Molecular Formula | C6H7KO2 |
| Molecular Weight (g/mol) | 150.22 |
| MDL Number | MFCD00016546 |
| InChI Key | CHHHXKFHOYLYRE-STWYSWDKSA-M |
| IUPAC Name | potassium (2E,4E)-hexa-2,4-dienoate |
| SMILES | [K+].C\C=C\C=C\C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605716
|
Spectrum Chemical Mfg Cor
P1408125GM |
125 g | Amber Glass Bottle |
Each for $327.13
|
|
||||
|
18605717
|
Spectrum Chemical Mfg Cor
P1408500GM |
500 g | Amber Glass Bottle |
Each for $710.37
|
|
||||
|
18605719
|
Spectrum Chemical Mfg Cor
P140812KG |
12 kg | Poly Pail |
Each for $4,690.13
|
|
||||
|
18605720
|
Spectrum Chemical Mfg Cor
P140845KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Potassium Sorbate, Powder, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 590-00-1 | |
| 150.22 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| [K+].C\C=C\C=C\C([O-])=O |
| C6H7KO2 | |
| MFCD00016546 | |
| potassium (2E,4E)-hexa-2,4-dienoate |
Specifications
| 590-00-1 | |
| 125 g | |
| MFCD00016546 | |
| [K+].C\C=C\C=C\C([O-])=O | |
| 150.22 | |
| FCC | |
| Amber Glass Bottle |
| 1 | |
| C6H7KO2 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| potassium (2E,4E)-hexa-2,4-dienoate | |
| 98 to 101% | |
| 0.01 |