missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sorbate, Powder, FCC, 98-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Potassium Sorbate, Powder, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 125 g |
| Percent Purity | 98 to 101% |
| Grade | FCC |
| Loss on Drying | 0.01 |
| Packaging | Amber Glass Bottle |