missing translation for 'onlineSavingsMsg'
Learn More

Medisca Potassium Sorbate NF

Catalog No. 14186039
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186039 500 g
14185502 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14186039 Supplier Medisca Supplier No. 083308
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Sorbate NF; white to off-white crystals or powder.

Potassium Sorbate NF; white to off-white crystals or powder.

Specifications

Chemical Name or Material Potassium Sorbate NF
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 24634-61-5
Type Drug Standard
Molecular Weight (g/mol) 150.22
Quantity 500 g
Percent Purity 98 to 102 % (Potassium Sorbate) (Dried Basis)
Grade NF