missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sorbate 99.0+%, TCI America™
$32.28 - $58.85
Chemical Identifiers
| CAS | 24634-61-5 |
|---|---|
| Molecular Formula | C6H7KO2 |
| Molecular Weight (g/mol) | 150.218 |
| MDL Number | MFCD00016546 |
| InChI Key | CHHHXKFHOYLYRE-STWYSWDKSA-M |
| Synonym | potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c |
| PubChem CID | 23676745 |
| ChEBI | CHEBI:77868 |
| IUPAC Name | potassium;(2E,4E)-hexa-2,4-dienoate |
| SMILES | CC=CC=CC(=O)[O-].[K+] |
Chemical Identifiers
| 24634-61-5 | |
| 150.218 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| 23676745 | |
| potassium;(2E,4E)-hexa-2,4-dienoate |
| C6H7KO2 | |
| MFCD00016546 | |
| potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c | |
| CHEBI:77868 | |
| CC=CC=CC(=O)[O-].[K+] |
Specifications
| 24634-61-5 | |
| C6H7KO2 | |
| potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c | |
| CC=CC=CC(=O)[O-].[K+] | |
| 150.218 | |
| CHEBI:77868 | |
| ≥99.0% (T) | |
| Potassium Sorbate [for Biochemical Research] |
| White-Yellow | |
| MFCD00016546 | |
| CHHHXKFHOYLYRE-STWYSWDKSA-M | |
| potassium;(2E,4E)-hexa-2,4-dienoate | |
| 23676745 | |
| 150.22 | |
| Granules |
Safety and Handling
EINECSNumber : (2)-1076
RTECSNumber : WG2170000
TSCA : Yes