missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sorbate 99.0+%, TCI America™

Supplier:  TCI America P195425G

Quantity 25 g

 View more versions of this product

Catalog No. P195425G


Only null left

Description

Description

Specifications

Chemical Identifiers

24634-61-5
150.218
CHHHXKFHOYLYRE-STWYSWDKSA-M
23676745
potassium;(2E,4E)-hexa-2,4-dienoate
C6H7KO2
MFCD00016546
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CHEBI:77868
CC=CC=CC(=O)[O-].[K+]

Specifications

Potassium Sorbate [for Biochemical Research]
White-Yellow
25 g
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CC=CC=CC(=O)[O-].[K+]
150.218
CHEBI:77868
≥99.0% (T)
24634-61-5
C6H7KO2
MFCD00016546
CHHHXKFHOYLYRE-STWYSWDKSA-M
potassium;(2E,4E)-hexa-2,4-dienoate
23676745
150.22
Granules
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1076

RTECSNumber : WG2170000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions