missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate, Tetrahydrate, Crystal, ACS, 99.0-102.0%, Spectrum™ Chemical
$308.09 - $11617.21
Chemical Identifiers
| CAS | 6381-59-5 |
|---|---|
| Molecular Formula | C4H12KNaO10 |
| Molecular Weight (g/mol) | 282.22 |
| InChI Key | VZOPRCCTKLAGPN-ZFJVMAEJSA-L |
| IUPAC Name | potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate |
| SMILES | O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18612895
|
Spectrum Chemical Mfg Cor
P1400125GM |
125 g | Poly Bottle |
Each for $308.09
|
|
||||
|
18612896
|
Spectrum Chemical Mfg Cor
P1400500GM |
500 g | Poly Bottle |
Each for $758.24
|
|
||||
|
18612897
|
Spectrum Chemical Mfg Cor
P140025KG |
2.5 kg | Poly Pail |
Each for $3,058.41
|
|
||||
|
18612898
|
Spectrum Chemical Mfg Cor
P140012KG |
12 kg | Poly Pail |
Each for $5,808.91
|
|
||||
|
18612899
|
Spectrum Chemical Mfg Cor
P140050KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 6381-59-5 | |
| 282.22 | |
| potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate |
| C4H12KNaO10 | |
| VZOPRCCTKLAGPN-ZFJVMAEJSA-L | |
| O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
Specifications
| 6381-59-5 | |
| 6.0 to 8.5 | |
| C4H12KNaO10 | |
| O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O | |
| 282.22 | |
| ACS |
| 1 | |
| 125 g | |
| VZOPRCCTKLAGPN-ZFJVMAEJSA-L | |
| potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate | |
| 99 to 102% | |
| Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.