missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate, Tetrahydrate, Crystal, ACS, 99.0-102.0%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P140025KG
Specifications
| 6381-59-5 | |
| 6.0 to 8.5 | |
| C4H12KNaO10 | |
| O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O | |
| 282.22 | |
| ACS |
| 1 | |
| 2.5 kg | |
| VZOPRCCTKLAGPN-ZFJVMAEJSA-L | |
| potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate | |
| 99 to 102% | |
| Poly Pail |
Chemical Identifiers
| 6381-59-5 | |
| 282.22 | |
| potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate |
| C4H12KNaO10 | |
| VZOPRCCTKLAGPN-ZFJVMAEJSA-L | |
| O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.