missing translation for 'onlineSavingsMsg'
Learn More

Potassium Pyrophosphate, Powder, Purified, Spectrum™ Chemical

$198.82 - $4774.03

Chemical Identifiers

CAS 7320-34-5
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612888
View Documents
Spectrum Chemical Mfg Cor
P1393500GM
500 g Poly Bottle
Each for $198.82
Only null left
 
18612889
View Documents
Spectrum Chemical Mfg Cor
P139325KG
2.5 kg Poly Bottle
Each for $545.18
Only null left
 
18612890
View Documents
Spectrum Chemical Mfg Cor
P139312KG
12 kg Poly Pail
Each for $1,648.74
Only null left
 
18612891
View Documents
Spectrum Chemical Mfg Cor
P139345KGBL
45 kg Fiber Drum
Each for $4,774.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7320-34-5

Specifications

7320-34-5
500 g
Poly Bottle
1
Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.