accessibility menu, dialog, popup

UserName

KKKKOPOOOPOOO

CAS RN 7320-34-5

IUPAC Name: tetrapotassium (phosphonatooxy)phosphonate
Synonyms: tetrapotassium pyrophosphate
Molecular Weight (g/mol): 330.33
Molecular Formula: K4O7P2
InChi Key: RYCLIXPGLDDLTM-UHFFFAOYSA-J
SMILES: [K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (4)
Physical Form 
  • (3)
  • (2)
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (7)
  • (7)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)