missing translation for 'onlineSavingsMsg'
Learn More

Potassium Propylxanthate 97.0+%, TCI America™

Catalog No. p053425g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P053425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P053425G Supplier TCI America Supplier No. P053425G
Only null left

Chemical Identifiers

CAS 2720-67-4
Molecular Formula C4H7KOS2
Molecular Weight (g/mol) 174.317
MDL Number MFCD00059687
InChI Key NZUFLKMNMAHESJ-UHFFFAOYSA-M
Synonym Propylxanthic Acid Potassium Salt, Potassium O-Propyl Dithiocarbonate
PubChem CID 23691005
IUPAC Name potassium;propoxymethanedithioate
SMILES CCCOC(=S)[S-].[K+]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 174.32
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Propylxanthate
EINECSNumber (2)-1787
RTECSNumber FG1581950
TSCA No